Ampicillin trihydrate, crystalline
Catalog # ICNA0219014725
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:403.45 g/mol
:GHS07
:GHS08
CAS Number:7177-48-2
Formula:C₁₆H₁₉N₃O₄S·3H₂O
MDL Number:MFCD00072036