Diethylenetriaminepentaacetic acid ≥90%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:67-43-6
:
Formula:C₁₄H₂₃N₃O₁₀
MDL Number:MFCD00004289
Melting Pt:219…220 °C (lit.)
Density:1.770 g/mL (lit.)
ADR:9,III
:393.35 g/mol
:GHS07
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Identity Test:Passes
- Purity:~90%
- FTIR:Conforms to Standard