Silicon dioxide
Catalog # ICNA0204660
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:25 kg
- Seq:4283
Specifications
About this item
:
:60.08 g/mol
Density:2.65 g/cm³ (20 °C)
Storage Temperature:Ambient
CAS Number:7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232