Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(−)-Apomorphine hydrochloride hydrate
(−)-Apomorphine hydrochloride hydrate ≥95%
(−)-Apomorphine hydrochloride hydrate ≥95%
:
Cayman Chemical
(−)-Apomorphine hydrochloride hydrate ≥95%
:
Cayman Chemical
:
58117-94-5
:
:
C₁₇H₁₇NO₂·HCl·xH₂O
:
303,8 g/mol
:
GHS08
:
GHS07
:
Freezer