ContactUsLinkComponent
Transport case for Self Contained Breathing Apparatus
Transport case for Self Contained Breathing Apparatus
Catalog # SCTS2014810
Supplier:  SCOTT SAFETY
Transport case for Self Contained Breathing Apparatus
Catalog # SCTS2014810
Supplier:  SCOTT SAFETY
Specifications
  • Description:
    Moulded carry case
  • Pk:
    1
Specifications
About this item
This carrying case made from moulded HDPE is used to hold one breathing apparatus set complete with face mask and cylinder.
  • Closed cell foam gasket
  • PE strap with velcro
The case features an internal insert to hold the apparatus, internal securing strap, sealing gasket, carrying handle and double latch fasteners.