Ion exchange resins, Diaion® UBK550 (Na)
46609.A1
:
- Particle Size:0,22 mm
- Pk:1 kg
- Description:Ion exchange fractionation resin, 8% cross-linked, strongly acidic gel type, 1,9 meq/ml on PS-DVB
These resins are ideal for use in chromatographic oligosaccharide sugar separations and sugar alcohol separations.