These seamless knitted liners with Intercept™ technology yarn either feature a ¾ dipped or a full nitrile coating.
- Excellent abrasion performance
- Good blade cut resistance
- Very good grip properties due to the nitrile micro finishing of the coating
- Flexible and comfortable
- Coating resistant to residual oils and greases present on handled parts
Intercept™: Specialised knitting technology which blends engineered, synthetic and natural fibers into high performance yarns. This provides high-cut protection with exceptional comfort and dexterity.
Fortix® Technology: Thin, resilient and breathable nitrile foam coating that dramatically extends the glove's working life and improves comfort in abrasive applications
Ansell Grip Technology™ provides a ‘Gecko-like’ grip in adverse conditions.
Ripel™ technology creates a liquid-impermeable layer in knit glove styles that prevents oil or lubricant from making even incidental contact with the wearer's skin.
Typical applications include: Assembling metal parts and components, aeronautics, assembly of white goods, finishing works, glass manufacturing, cutting small dry or lightly oiled parts, plastics injection and moulding, maintenance.
- Catalog No:
- ANSE11-947/10
- ANSE11-947/11
- ANSE11-948/8
- ANSE11-947/8
- ANSE11-947/9
- ANSE11-948/7
- ANSE11-948/9
- ANSE11-948/10
- ANSE11-948/11
- ANSE11-947/7
- Model:
- 11-947
- 11-947
- 11-948
- 11-947
- 11-947
- 11-948
- 11-948
- 11-948
- 11-948
- 11-947
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Liner material:
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Intercept™ yarn
- Finish:
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- ¾ dipped
- Gauge:
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Safety cuff
- Knit wrist
- Knit wrist
- Safety cuff
- Safety cuff
- Safety cuff
- Safety cuff
- Knit wrist
- EN norm:
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)
- EN 388:2016 (4X42B)