Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium chloride
Sodium chloride 0,9%, Ecotainer®
Sodium chloride 0,9%, Ecotainer®
Sodium chloride 0,9%, Ecotainer®
Supplier:
B. Braun
Certificates
Expand image
Sodium chloride 0,9%, Ecotainer®
Supplier:
B. Braun
Certificates
About this item
CAS Number:
7647-14-5
:
Formula:
NaCl
MDL Number:
MFCD00003477
:
58.44 g/mol
Related Information
Documentation
Certificates
Search by Lot Number
Documentation