PEEK tubing elbows
Supplier: Restek
Certificates
About this item
Tubing elbows are ideal for routing 1,6 mm PEEK tubing that snap the tubing into the elbow. Its prevent pinching of PEEK tubing, which can cause high pressure.
Tubing elbows are ideal for routing 1,6 mm PEEK tubing that snap the tubing into the elbow. Its prevent pinching of PEEK tubing, which can cause high pressure.