Anti-NKX2-2 Monoclonal Antibody [clone: NX2/1422R]
Catalog # ABNORAB00284
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:NK2 homeobox 2
- Antigen symbol:NKX2-2
- Clonality:Monoclonal
- Clone:NX2/1422R
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Isotype:IgG kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:4821
- Storage buffer:In 10 mM PBS.
- Storage temperature:Store at 4° C. For long term storage store at -20° C.
- Immunogen:Original antibody is raised against recombinant NKX2-2.
- Purification:Protein A purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal antibody raised against human NKX2-2.
Type: Primary
Antigen: NKX2-2
Clonality: Monoclonal
Clone: NX2/1422R
Conjugation: Unconjugated
Epitope:
Host:
Isotype: IgG, kappa
Reactivity: Human