Anti-IGHG4 Mouse Monoclonal Antibody [clone: IGHG4/1345]
ABNOMAB14563
: Abnova
- Antibody type:Primary
- Antigen name:IGHG4 Recombinant
- Antigen symbol:IGHG4
- Clonality:Monoclonal
- Clone:IGHG4/1345
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:3503
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Storage temperature:Store at 4° C.
- Immunogen:Recombinant protein corresponding to full length human IGHG4.
- Purification:Protein A/G purification
- Size:100 µg
- Pk:100 µG
Mouse monoclonal antibody raised against full length recombinant human IGHG4.
Type: Primary
Antigen: IGHG4
Clonality: Monoclonal
Clone: IGHG4/1345
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human