Order Entry
Germany
ContactUsLinkComponent
 
Economy water vapor trap
Catalog # 23988.
Supplier:  Merck
Economy water vapor trap
Catalog # 23988.
Supplier:  Merck

Specifications

  • Description:
    Economy moisture trap, 400 ml, fitting size 1/4"
  • Pk:
    1

Specifications

About this item

This economy water vapor trap is designed for moisture removal for compressed air service to pneumatically-controlled devices.

  • Purifier body made of clear polycarbonate
  • Trap contains a mixture of Molecular Sieves 4A and 13X
  • Not to be used with carrier gases due to permeability of purifier body