Order Entry
Germany
ContactUsLinkComponent
Labels for LABXPERT™ and IDXPERT™ printers, type B-499
Labels for LABXPERT™ and IDXPERT™ printers, type B-499
Catalog # BRDY60913
Supplier:  Brady
undefined
Labels for LABXPERT™ and IDXPERT™ printers, type B-499
Catalog # BRDY60913
Supplier:  Brady

Specifications

  • Type:
    B-499
  • Colour:
    Black/white
  • Description:
    Printer labels
  • Dimensions:
    13×25 mm
  • Material:
    Nylon
  • Seq:
    11
  • Packed:
    225 labels/cartridge
  • Pk:
    1
  • Adhesive:
    Permanent acrylic

Specifications

About this item

Printable nylon cloth labels for LABXPERT™ and IDXPERT™ printers.

  • Die-cut labels are supplied on a roll
  • Contains some adhesive backed wire marking materials, and also labels that are pre-printed with headers or templates
  • Supplied with the ribbon seperate, not in the same enclosure as the tape/sleeves

This material is UL recognised with its respective ribbon. The recommended ribbon series are available for the appropriate ribbon to use. Certain auto-formatting features may not be enabled when using this part in an IDXPERT™ printer.