HPLC column tubing
Supplier: Restek
Certificates
About this item
Manufactured from fine, chromatographic grade 316 stainless steel.
- Pre polished tubing ensures burr-free cut
- No additional treatment needed
Corrosion resistant, ultrasonically cleaned, and passivated.