Orders LinkContactUsLinkComponent
Dual canister sampling manifolds, stainless steel/Siltek® treated
Dual canister sampling manifolds, stainless steel/Siltek® treated
Supplier:   Restek
Dual canister sampling manifolds, stainless steel/Siltek® treated
Supplier:  Restek
Certificates
Dual canister sampling manifolds, stainless steel/Siltek® treated
Dual canister sampling manifolds, stainless steel/Siltek® treated
2 Options Available Below
Product Details & Documents
Dual canister sampling manifold minimises sampling variability through a single sample inlet and flow controller by which the sample is evenly collected between two canisters. Use of a single flow controller eliminates flow rate variability, as well as environmental variables common with collocated samples. It may also be used with two flow controllers to monitor individual canister vacuum.
  • Precise dimensions (W×H: 241,3×88,9 mm) provide accurate splitting of sample between two canisters
  • Minimises leaks
  • Thick-walled stainless steel tubing is rugged enough for field use
  • Uses 6,35 mm compression connections
Caution: Do not use this manifold as a handle to pick up 2 canisters.
Product Family Options
Product Information
  • Availability
    Price
  • REST24998
    Dual canister sampling manifold, stainless steel
    Dual canister sampling manifold, stainless steel
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 355,74 €
    355,74 €
    1 * 1 ST
       355,74 €
  • REST24999
    Dual canister sampling manifold, Siltek® treated
    Dual canister sampling manifold, Siltek® treated
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 454,38 €
    454,38 €
    1 * 1 ST
       454,38 €