Order Entry
Germany
ContactUsLinkComponent
VWR® Facial Tissues
VWR® Facial Tissues
  115-0600
undefined
VWR® Facial Tissues
  115-0600

 

  • Packed:
    150 sheets per box
  • L×W:
    195×200 mm
  • Pk:
    150

 

 

Two ply facial tissues made from 100% cellulose.

  • Extra soft.

Colour: White