Orders LinkContactUsLinkComponent
Vacuum gauges
Vacuum gauges
Supplier:   Restek
Vacuum gauges
Supplier:  Restek
Certificates
Vacuum gauges
2 Options Available Below
Product Details & Documents
These vacuum gauges are made of stainless steel wetted surfaces with rear mount.
Suitable for passive air sampling kits.
Product Family Options
Product Information
  • Availability
    Price
  • REST24269
    Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 3,175 mm NTP fitting
    Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 3,175 mm NTP fitting
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,38 €
    199,38 €
    1 * 1 ST
       199,38 €
  • REST24270
    Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 6,35 mm NTP fitting
    Vacuum gauge, −30 to 0 Hg, Ø 51 mm, 6,35 mm NTP fitting
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 199,38 €
    199,38 €
    1 * 1 ST
       199,38 €