Order Entry
Germany
ContactUsLinkComponent
Secondary columns for GCxGC (fused silica), Rxi-17Sil
Secondary columns for GCxGC (fused silica), Rxi-17Sil
Catalog # REST15116
Supplier:  Restek
undefined
Secondary columns for GCxGC (fused silica), Rxi-17Sil
Catalog # REST15116
Supplier:  Restek

Specifications

  • Column Format:
    Capillary
  • Film thickness:
    0,18 µm
  • Ø int.:
    0,18 mm
  • Length:
    2 m
  • Phase:
    Rxi®-17Sil MS
  • Pk:
    1

Specifications

About this item

Convenient 2 m length is ideal for use as a secondary column in GCxGC analyses.

  • Midpolarity Crossbond phase
  • Excellent inertness and selectivity for active environmental compounds, such as PAHs
  • 340/360 °C upper temperature limits
  • Low-bleed for use with sensitive detectors, such as MS