Order Entry
Germany
ContactUsLinkComponent
Staurosporine (from Streptomyces species), InSolution™, Millipore®
Staurosporine (from Streptomyces species), InSolution™, Millipore®
Catalog # 569396-100
CAS Number:  
Staurosporine (from Streptomyces species), InSolution™, Millipore®
Catalog # 569396-100
CAS Number:  

Specifications

  • Pk:
    100 µG
  • Seq:
    1286

Specifications

About this item

 :
 :466.541 g/mol
CAS Number:62996-74-1
Formula:C₂₈H₂₆N₄O₃
MDL Number:MFCD18252446