ContactUsLinkComponent
Capillary inlet adaptor fitting kit
Capillary inlet adaptor fitting kit
Catalog # REST27184
Supplier:  Restek
undefined
Capillary inlet adaptor fitting kit
Catalog # REST27184
Supplier:  Restek
Specifications
  • Description:
    Capillary inlet adaptor fitting kit, 1/16"
  • Pk:
    1
Specifications
About this item
This capillary inlet adaptor fitting kit is ideal for split/splitless fitting for capillary columns.
  • Split/splitless fitting that accepts standard capillary ferrules
  • Easier to install capillary columns because the nut protrudes farther from the insulated injection port chamber
Delivery: Kit includes adaptor fitting, capillary nut, gold-plated 0,8 mm ID dual Vespel® ring inlet seal, and one 0,4 mm ID ferrule.