ContactUsLinkComponent
Septum nut removal tool
  REST24918
 :  Restek
undefined
Septum nut removal tool
  REST24918
 :  Restek
  • Description:
    Septum nut removal tool
  • Pk:
    1
This septum nut removal tool is ideal for Agilent 5890/6850/6890/7890 GCs and it easily removes the septum nut without touching the heated nut.
  • Unique, ergonomic handle and easy to grip
  • Nut remains in tool for quick reattachment