ContactUsLinkComponent
Benchtop turbidimeters, TU52
Benchtop turbidimeters, TU52
Supplier:  Hach
Certificates
undefined
Benchtop turbidimeters, TU52
Supplier:  Hach
Specifications
  • Catalog No:
  • HACHLZY898
  • HACHLPV4429903022
  • HACHLZY901
  • HACHLZY837
  • HACHLZY835
  • HACHLZY899
  • HACHLZY877
  • Light source:
  • Up to 10 mNTU
  • Accuracy:
  • ±2,0 % +0,01 NTU (0 - 40 NTU) ; ±10 % (40 - 1000 NTU based on Formazin primary standard @ 25 °C)
  • Display:
  • Colour LCD with touch screen
  • Repeatability:
  • Up to 40 NTU: Better than 1,0 % or ±0,002 NTU on Formazin @ 25 °C (77 °F)
    Over 40 NTU: Better than 3,5 % on Formazin @ 25 °C (77 °F)
  • Resolution:
  • 0,0001 NTU ; FNU ; TE/F ; FTU ; EBC ; mg/L
  • Power requirements:
  • 50 - 60 Hz/100 - 240 VAC