Septa holder kit
Catalog # REST21299
Supplier: Restek
Specifications
- Description:Septa holder kit with septum support and holder
- Pk:1
Specifications
About this item
This kit can be used for Thermo Scientific TRACE, 8000, 8000 TOP and Focus SSL. It is made from high quality stainless steel.
Delivery: Includes septum support and holder.