Connectors, Vu2 Union®
: Restek
These connectors can be used to connect a guard column to an analytical column, a column to a transfer line, two columns in series or repair a broken column.
- Open design of the connector allows visual confirmation of the seal
- Secondary seals ensure leak-tight connections
A Press-Tight® union in the Vu2 Union® connector joins the fused silica tubing ends together; the ferrule and knurled nut at each end of the connector hold the tubing in place via a secondary seal between the ferrule and the Press-Tight® union. Each knurled nut applies independent pressure to each ferrule to make leak-tight seals with the column ends.
: Kit includes Vu2 Union® body, two knurled nuts, two Press-Tight® unions and four ferrules.