Order Entry
Germany
ContactUsLinkComponent
Tube cutting machine, SSI TC-20
Tube cutting machine, SSI TC-20
Catalog # REST23029
Supplier:  Restek
undefined
Tube cutting machine, SSI TC-20
Catalog # REST23029
Supplier:  Restek

Specifications

  • Description:
    SSI tubing cutter machine, 203×159×108 mm, 5 kg
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

This tube cutting maching cuts ¹/₁₆", ¹/₈" or ¹/₄" tubing with inside diameter as small as 0,008".

  • Electrically operated bench-top model
  • Handy dressing tool on the swing arm removes burrs and reams tubing
  • Voltage selectable 110 to 120/220 to 240 volts, 50 to 60 Hz