ContactUsLinkComponent
Rough pump oil
Catalog # REST22687
Supplier:  Restek
undefined
Rough pump oil
Catalog # REST22687
Supplier:  Restek
Specifications
  • Description:
    Rough pump oil #19, for MSD pumps
  • Pk:
    1 Bottle
Specifications
About this item
This rough pump oil ideal for MSD and oil vacuum pumps is formulated from crude oil stocks known for their durability and line-lubricating qualities.
It can be used in Agilent 5973/5972/5971 and GCD mass spec systems or in other MSD systems that require rough pump oil.