Protease inhibitor cocktail, MP Biomedicals
Catalog # ICNA0215883701
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 KIT
- Description:Protease inhibitor cocktail
Specifications
About this item
A broad spectrum protease inhibitor cocktail which covers both endopeptidases and exopeptidases, non-specific and specific. The cocktail consists of four major protease inhibitors such as AEBSF, Sodium EDTA, Leupeptin and Pepstain A.