ContactUsLinkComponent
Mob caps, non woven PP
Mob caps, non woven PP
Catalog # HOIT109758
undefined
Mob caps, non woven PP
Catalog # HOIT109758
Specifications
  • Ø:
    530 mm
  • Garment application:
    Clinical,
    Food
  • Colour:
    White
  • Seq:
    03
  • Pk:
    1.000
Specifications
About this item
Disposable mob caps made from non woven PP.
  • Lightweight, breathable and comfortable to wear
  • Food contact approved
Ideal for use in hospitals, food processing, manufacturing and other general applications.