Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(2'Z,3'E)-6-Bromoindirubin-3'-oxime
(2'Z,3'E)-6-Bromoindirubin-3'-oxime, Millipore®
(2'Z,3'E)-6-Bromoindirubin-3'-oxime, Millipore®
(2'Z,3'E)-6-Bromoindirubin-3'-oxime, Millipore®
Supplier:
Merck Millipore (Calbiochem)
Certificates
Expand image
(2'Z,3'E)-6-Bromoindirubin-3'-oxime, Millipore®
Supplier:
Merck Millipore (Calbiochem)
About this item
CAS Number:
667463-62-9
:
Formula:
C₁₆H₁₀BrN₃O₂
MDL Number:
MFCD08705318
:
356.18 g/mol