Order Entry
Germany
ContactUsLinkComponent
Anti-CD16 / CD32 Rat Monoclonal Antibody (PE) [clone: 2.4G2]
Anti-CD16 / CD32 Rat Monoclonal Antibody (PE) [clone: 2.4G2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD16 / CD32 Rat Monoclonal Antibody (PE) [clone: 2.4G2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0161-U025
  • 50-0161-U100
  • Antigen symbol:
  • FCGR3, IGFR3, FCGR2, IGFR2, FC Receptor
  • FCGR3, IGFR3, FCGR2, IGFR2, FC Receptor
  • Antigen name:
  • CD16 / CD32
  • CD16 / CD32
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • 2.4G2
  • 2.4G2
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 14130/14131
  • 14130/14131
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • 2-8°C protected from light
  • 2-8°C protected from light

Specifications

Related Information