Tubing, stainless steel 304, coiled welded/drawn
Supplier: Restek
Certificates
About this item
These Sulfinert® treated tubings are recommended for chromatography applications, gas delivery systems, lower pressures and inert applications.
- Stainless steel fabrication
- Maximum temperature of 450 °C in an inert atmosphere
Sulfinert® is a required treatment for metal components when analysing for parts-per-billion levels of organo-sulphur compounds.