Order Entry
Germany
ContactUsLinkComponent
 
Klenow (3'-5' exo-) (from E. coli carrying the Klenow (3'-5' exo-) gene)
  ENZOENZ44001U020
 :  
Klenow (3'-5' exo-) (from E. coli carrying the Klenow (3'-5' exo-) gene)
  ENZOENZ44001U020
 :  

 

  • Pk:
    20 µl
  • Source:
    E. coli carrying the Klenow (3'-5' exo-) gene
  • Seq:
    02
  • Storage conditions:
    Stable for at least 1 year after receipt when stored at –20 °C
  • Form:
    Liquid
  • Shipping conditions:
    Dry Ice
  • Enzyme name:
    Klenow (3'-5' exo-)
  • Enzyme activity:
    10000 U/mg
  • Purity:
    ≥99% (SDS-PAGE)
  • Formulation:
    In 20 mM TRIS-HCl, pH 7,5, containing 50% glycerol, 0,1 mM EDTA and 1 mM DTT.

 

 

Klenow (3′-5′ exo-) is a mesophilic DNA polymerase deficient in both proofreading (3′-5′) and nick-translation (5′-3′) nuclease activities, and that displays a moderate strand displacement activity during DNA synthesis. The protein is expressed as a truncated product of the E.coli PolA gene and contains the D355A and E357A mutations.

  • Used for DNA labeling
  • Not for 5´→3´ or 3´→5´exonuclease activity
  • N-terminal truncated DNA Polymerase I which retains polymerase activity

It is formulated of liquid in 20 mM TRIS-HCl, pH 7,5, containing 50% glycerol, 0,1 mM EDTA and 1 mM DTT.