PLOT column particle traps
Supplier: Restek
Certificates
About this item
When extreme pressure shocks and gas flow changes are anticipated there is a possibility for particles to dislodge. In those cases, using particle traps is recommended.
- Protects detector and valves; connects between column and detector or valve
- Eliminates detector spikes and scratches in valve rotors
Delivery: Includes two press-tight connectors, a 2,5 m column, and high-temperature string.