Order Entry
Germany
ContactUsLinkComponent
PLOT columns, Rt®-QS-BOND columns (fused silica)
PLOT columns, Rt®-QS-BOND columns (fused silica)
  REST19740
 :  Restek
undefined
PLOT columns, Rt®-QS-BOND columns (fused silica)
  REST19740
 :  Restek

 

  • Column Format:
    Capillary
  • Film thickness:
    10,00 µm
  • Ø int.:
    0,32 mm
  • Length:
    30 m
  • Pk:
    1

 

 

Intermediate polar, porous divinylbenzene homopolymer.

  • Similar phase: GS-Q
  • Separates ethane, ethylene, and acetylene to baseline