ContactUsLinkComponent
VWR® Rota® Tri, pH indicator, 1 to 11
VWR® Rota® Tri, pH indicator, 1 to 11
Catalog # 85409.600
Supplier:  VWR Chemicals
undefined
VWR® Rota® Tri, pH indicator, 1 to 11
Catalog # 85409.600
Supplier:  VWR Chemicals
Specifications
  • pH gradation:
    1 · 2 · 3 · 4 · 5 · 6 · 7 · 8 · 9 · 10 · 11
  • pH range:
    1 - 11
  • Pk:
    5
Specifications
About this item
These tri indicator papers show three different colours for each pH-value.
  • High precision due to the 3 colour scale
  • Easy to use
  • Produced from high quality paper to give good colour matching of paper to printed calibration scale
Tri paper has a hydrophobic barrier between the different indicators which ensures a distinctive separation of the colour bands.
Delivery: Supplied in plactic reels that ensure protection and long term stability. Reel of 6 m length, width: 14 mm