Rat recombinant Hip (from E. coli)
100 µG
Catalog # BMOLSPP-767E
Supplier: BIOMOL RESEARCH LABORATORIES
Rat recombinant Hip (from <i>E. coli</i>)
Catalog # BMOLSPP-767E
Supplier: BIOMOL RESEARCH LABORATORIES
CAS Number:
Jump to:
Product Details & Documents
Hsc70-interacting protein (Hip) is a cytosolic protein that exists as a homo-oligomer and participates in the regulation of the heat shock protein Hsc70 in eukaryotic cells.
- Produced in E. coli
- Purified by multi-step chromatography (SDS-PAGE, western blot)
It has a liquid formulation in 20 mM MOPS at pH 7,2, containing 50 mM sodium chloride and 2 mM magnesium chloride.
Specifications
Specifications
- Pk:100 µG
- Cat. No.:BMOLSPP-767E
- Protein/peptide type:Recombinant
- Protein/peptide name:Hip
- Storage conditions:–80 °C
- Formulation:Liquid
- Protein synonyms:HSC70-interacting protein
- Purity:>85%
- pH:7,2
- Tested applications:Western blot
- Species:Rat
- Source:E. coli
- Molecular weight:~54 kDa
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



