ContactUsLinkComponent
Solid phase extraction cartridges, CarboPrep®
Solid phase extraction cartridges, CarboPrep®
Catalog # REST26206
Supplier:  Restek
undefined
Solid phase extraction cartridges, CarboPrep®
Catalog # REST26206
Supplier:  Restek
Specifications
  • Column Format:
    Cartridge
  • Description:
    CarboPrep 200 reversing cartridge
  • Bed weight:
    500 mg
  • Pk:
    30
Specifications
About this item
The reversing cartridge design allows convenient inverted elution of strongly retained analytes using minimum solvent volumes. Ideal for extraction of pesticides in water.
  • High adsorbent capacity (surface area ~200 m²/g) for large volume sampling
Chromatographic grade graphitised carbon provides consistent and quantitative recoveries of a wide variety of semivolatiles, pesticides and herbicides.