Orders LinkContactUsLinkComponent
<i>Clostridium difficile</i> toxoid B
Clostridium difficile toxoid B
Clostridium difficile toxoid B, 50 µg, 50 µG
Catalog # ENZOBMLG1550050
Supplier:   Enzo Life Sciences
<i>Clostridium difficile</i> toxoid B
Catalog # ENZOBMLG1550050
Supplier:  Enzo Life Sciences
CAS Number:  
Clostridium difficile toxoid B
Clostridium difficile toxoid B, 50 µg, 50 µG
CAS Number:  
Price
1 180,00 €
Log in for availability
1 * 50 µG
Product Details & Documents
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
  • Minimal lot-to-lot variation
  • Inactivated by formaldehyde
  • Lyophilised for ease of handling and long term stability
Specifications
Specifications
  • Pk:
    50 µG
  • Description:
    Clostridium difficile toxoid B, 50 µg