Total organic carbon (TOC) vial
Catalog # IONICHMI90606
Supplier: IONICS
Specifications
- Cap included:Yes
- Capacity:40,00 ml
- Description:TOC vial, with cap and septa
- Pk:72
Specifications
About this item
TOC vials are designed for use in critical pharmaceutical applications such as USP, EP, JP and KP water testing.