Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Lubricants
Grease, silicone
Grease, silicone
Grease, silicone
Catalog # 1.07746.0100
Supplier:
Merck
SDS
Expand image
Grease, silicone
Catalog # 1.07746.0100
Supplier:
Merck
SDS
Specifications
Description:
Grease, silicone, density: 1 g/cm
3
(20 °C), flash point >300 °C
Pk:
100 g
Specifications
About this item
Product Details
Documentation
Product Details
Documentation