Reverse frits
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
About this item
Ideal for the filtration of substances sensitive to oxidation and hydrolysis. With filter plate, one-way cock and two standard ground-nuts.
2 Options Available Below
Product Details & Documents
Ideal for the filtration of substances sensitive to oxidation and hydrolysis. With filter plate, one-way cock and two standard ground-nuts.
Product Family Options
Product Information
- Frit materialFrit volumeFrit Ø ext.AvailabilityPrice
- NEUB4322-0100-144Reverse frit with glass stopper P4Reverse frit with glass stopper P4Borosilicate glass 3.3100 ml30 mmLog in for availability1 * 1 ST360,00 €
- NEUB4322-0100-294Reverse frit with glass stopper P4Reverse frit with glass stopper P4Borosilicate glass 3.3100 ml30 mmLog in for availability1 * 1 ST311,00 €