Lysonase
Supplier: Merck Millipore (Novagen)
Certificates
About this item
Lysonase™ bioprocessing reagent is an optimised, ready-to-use convenient blend of rLysozyme™ solution and benzonase nuclease which is ideal for protein extraction.
rLysozyme solution contains a highly purified and stabilized recombinant lysozyme with specific activity 250 times greater than that of chicken egg white lysozyme. Benzonase nuclease is a genetically engineered nonspecific endonuclease that degrades all forms of DNA and RNA (single stranded, double stranded, circular, linear), reducing extract viscosity and increasing protein yield. The combined activities of rLysozyme and benzonase nuclease significantly increase protein extraction efficiency and facilitate downstream processing of protein extracts.
Specifications
- Enzyme name:Lysonase