DNA ladder, O'RangeRuler™
Supplier: Thermo Fisher Scientific
Certificates
About this item
O'RangeRuler™ DNA ladders are step ladders that consist of basic unit repeats of 10, 15, 20, 50, 100, 200, or 500 bp. The ladders have a higher density of bands at a specific DNA size range and are therefore suitable for more accurate sizing of DNA in agarose or non denaturing polyacrylamide gels.
- Step ladders - ligation products of DNA fragments of equal increments
- Sharp bands, evenly spaced reference bands
- Ready to use - premixed with loading dye for direct loading
- Room temperature storage
- Supplied with loading dye for sample DNA
Delivery: Supplied with 6X orange DNA loading dye for sample DNA.