GC columns, Stabilwax-MS (fused silica)
: Restek
Stabilwax-MS columns ensure reproducible retention times from run to run, even with temperature cycling. When methods require trace analysis, this highly polar, low-bleed stationary phase produces excellent signal-to-noise levels. Ideal for food and flavor analysis (e.g., essential oils), fragrance and allergen analysis, as well as industrial solvent and chemical analysis.
- High-polarity, stable polyethylene glycol (PEG) stationary phase
- Low bleed and rugged enough to withstand repeated temperature cycles without retention time shifting
- Temperature range: 40 to 250/260 °C
- Equivalent to USP G14, G15, G16, G20 and G39 phases
Frequently Bought Together