ContactUsLinkComponent
Adhesion promoter, Silane A 174
Adhesion promoter, Silane A 174
Catalog # 1.07673.0050
Supplier:  Merck
Adhesion promoter, Silane A 174
Catalog # 1.07673.0050
Supplier:  Merck
Specifications
  • Packed:
    50 ml glass bottle
  • Description:
    Silane A 174 adhesion promoter
  • Flash point:
    108 °C
  • Pk:
    50 mL
Specifications
About this item
Silane A 174 (also known as Polyfix® 100, methacryloxypropyl trimethoxysilane) is an adhesion promoter. Has a molar mass 248,35 g/mol.