ContactUsLinkComponent
Tubing cutter, Clean-Cut
Tubing cutter, Clean-Cut
Catalog # REST25069
Supplier:  Restek
undefined
Tubing cutter, Clean-Cut
Catalog # REST25069
Supplier:  Restek
Specifications
  • Description:
    Clean-cut tubing cutter
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This tubing cutter provides burr-free, perpendicular cuts that will not distort the tubing O.D. or close the I.D.
Use for PEEK, PTFE, Tefzel® and other polymeric tubing.