Pronase E (from Streptomyces griseus)
Supplier: Merck
Certificates
About this item
Pronase E is a mixture of proteolytic enzymes that is obtained from Streptomyces griseus. It can be used in the in diagnostics in histopathology as it hydrolyses proteins into individual amino acids.
Specifications
- CAS No.:9036-06-0
- Source:Streptomyces griseus
- Storage conditions:2...8 °C
- Form:Slightly brown
- Enzyme name:Pronase E
- Enzyme activity:≥4000000 u/g
- pH:6 - 7