Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silicone oil
Silicone oil, 100 - 300 mPa.s (20 °C) for oil baths, Supelco®
Silicone oil, 100 - 300 mPa.s (20 °C) for oil baths, Supelco®
:
Merck
Silicone oil, 100 - 300 mPa.s (20 °C) for oil baths, Supelco®
:
Merck
:
68083-14-7
:
63148-62-9
:
MFCD00132673
:
>315 °C (1013 hPa)
:
1.04 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
>295 °C
msds_107742