Orders LinkContactUsLinkComponent
Trays, SteriWare®
Trays, SteriWare®
Supplier:   Sampling Systems
Trays, SteriWare®
Supplier:  Sampling Systems
Certificates
Trays, SteriWare®
Trays, SteriWare®
2 Options Available Below
Product Details & Documents
Trays made of PP, translucent.
  • Assembled and packed in a cleanroom
  • Individually bagged
  • FDA & EU 10/2011 conforming materials
  • BSE/TSE free
  • Full batch traceability
  • Available sterile (gamma irradiated)
The disposable tray is ideal for hygienic applications within hospitals and pharmaceutical factories.
Product Family Options
Product Information
  • Capacity
    W×D×H
    Availability
    Price
  • SAMP8081P-3001
    Non sterile
    Non sterile
    3000 ml
    265×310×55 mm
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 350,00 €
    350,00 €
    1 * 30 ST
       350,00 €
  • Sterile
    3000 ml
    265×310×55 mm
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 477,00 €
    477,00 €
    1 * 30 ST
       477,00 €