Internal reducing union, zero dead volume, Valco®
Supplier: Restek
Certificates
About this item
Internal reducing union used to connect two sizes of tubing, using zero-dead-volume fittings on each end. Suitable for 1,6 to 3,2 mm OD tubing.
Delivery: Supplied with stainless steel ferrules.